Assay Details
| Gene Symbol: SUMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035458.1 | NP_001030535.1 | 3 - 4 | 263 | 87 | |
| GenBank mRNA | BC102884.1 | - | 3 - 4 | 254 | 87 | |
| BT020989.1 | - | 3 - 4 | 263 | 87 | ||
More Information
Set Membership: |
|

SUMO1
Genomic Map