Assay Details
Gene Symbol: PTPN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040576.1 | NP_001035666.1 | 2 - 3 | 506 | 66 | |
GenBank mRNA | BC149270.1 | - | 4 - 5 | 643 | 66 | |
BT021722.1 | - | 2 - 3 | 506 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
