Assay Details
Gene Symbol: SLC35B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076965.1 | NP_001070433.1 | 1 - 2 | 98 | 116 | |
GenBank mRNA | BC103464.1 | - | 1 - 2 | 98 | 116 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC103464.1, NM_001076965.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
