Assay Details
| Gene Symbol: GPR89A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076970.1 | NP_001070438.1 | 11 - 12 | 1141 | 133 | |
| GenBank mRNA | BC120025.1 | - | 12 - 13 | 1241 | 133 | |
| BT021195.1 | - | 11 - 12 | 1141 | 133 | ||
| BT021606.1 | - | 11 - 12 | 1137 | 133 | ||
More Information
Set Membership: |
|

GPR89A
Genomic Map