Assay Details
| Gene Symbol: RHOB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077922.1 | NP_001071390.1 | 1 | 2048 | 154 | |
| GenBank mRNA | BC103067.1 | - | 1 | 2048 | 154 | |
More Information
Set Membership: |
|

RHOB
Genomic Map