Assay Details
| Gene Symbol: MMAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079632.1 | NP_001073100.1 | 1 - 2 | 171 | 56 | |
| GenBank mRNA | BC111194.1 | - | 1 - 2 | 122 | 56 | |
| BT021748.1 | - | 1 - 2 | 171 | 56 | ||
More Information
Set Membership: |
|

MMAB
Genomic Map