Assay Details
Gene Symbol: AIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173985.2 | NP_776410.1 | 3 - 4 | 254 | 69 | |
GenBank mRNA | AF348450.1 | - | 3 - 4 | 254 | 69 | |
BC102890.1 | - | 3 - 4 | 243 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
