Assay Details
Gene Symbol: CD69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174014.2 | NP_776439.1 | 4 - 5 | 568 | 68 | |
GenBank mRNA | AF272828.1 | - | 4 - 5 | 568 | 68 | |
BC142335.1 | - | 4 - 5 | 576 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
