Assay Details
| Gene Symbol: LDHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316338.1 | NP_001303267.1 | 2 - 3 | 275 | 63 | |
| NM_174100.2 | NP_776525.2 | 2 - 3 | 275 | 63 | ||
| GenBank mRNA | AJ401268.1 | - | 2 - 3 | 239 | 63 | |
| BC102217.1 | - | 2 - 3 | 206 | 63 | ||
| BC142006.1 | - | 2 - 3 | 276 | 63 | ||
| BC151427.1 | - | 2 - 3 | 206 | 63 | ||
| BT021009.1 | - | 2 - 3 | 243 | 63 | ||
| DT720873.1 | - | 2 - 3 | 216 | 63 | ||
More Information
Set Membership: |
|

LDHB
Genomic Map