Assay Details
Gene Symbol: KLRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174376.2 | NP_776801.1 | 2 - 3 | 258 | 114 | |
GenBank mRNA | AY075101.1 | - | 2 - 3 | 258 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() |
