Assay Details
Gene Symbol: KLRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174376.2 | NP_776801.1 | 4 - 5 | 590 | 141 | |
GenBank mRNA | AY075101.1 | - | 4 - 5 | 590 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() |
