Assay Details
| Gene Symbol: LECT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174380.2 | NP_776805.1 | 1 - 2 | 46 | 73 | |
| GenBank mRNA | AB001350.1 | - | 1 - 2 | 46 | 73 | |
| BC103166.1 | - | 1 - 2 | 221 | 73 | ||
| D89011.1 | - | 1 - 2 | 176 | 73 | ||
More Information
Set Membership: |
|

LECT2
Genomic Map