Assay Details
Gene Symbol: PTGES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174443.2 | NP_776868.1 | 1 - 2 | 139 | 97 | |
GenBank mRNA | AY032727.1 | - | 1 - 2 | 139 | 97 | |
BC120228.1 | - | 1 - 2 | 168 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
