Assay Details
| Gene Symbol: VEGFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174487.2 | NP_776912.1 | 2 - 3 | 201 | 71 | |
| GenBank mRNA | AB004273.1 | - | 2 - 3 | 201 | 71 | |
| AB004274.1 | - | 2 - 3 | 66 | 71 | ||
More Information
Set Membership: |
|

VEGFB
Genomic Map