Assay Details
Gene Symbol: YWHAE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174491.3 | NP_776916.1 | 4 - 5 | 668 | 117 | |
GenBank mRNA | AF043735.1 | - | 4 - 5 | 615 | 117 | |
BC102928.1 | - | 4 - 5 | 668 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
