Assay Details
| Gene Symbol: PLA2G15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174560.2 | NP_776985.2 | 2 - 3 | 393 | 69 | |
| GenBank mRNA | AY072914.1 | - | 2 - 3 | 266 | 69 | |
| BC149389.1 | - | 2 - 3 | 356 | 69 | ||
| BT021838.1 | - | 2 - 3 | 393 | 69 | ||
More Information
Set Membership: |
|

PLA2G15
Genomic Map