Assay Details
| Gene Symbol: PRSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174690.1 | NP_777115.1 | 3 - 4 | 458 | 128 | |
| GenBank mRNA | BC134794.1 | - | 3 - 4 | 472 | 128 | |
| BC142036.1 | - | 3 - 4 | 472 | 128 | ||
More Information
Set Membership: |
|

PRSS2
Genomic Map