Assay Details
| Gene Symbol: HSP90B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174700.2 | NP_777125.1 | 4 - 5 | 508 | 68 | |
| GenBank mRNA | AB025193.1 | - | 4 - 5 | 508 | 68 | |
| BC104549.1 | - | 4 - 5 | 459 | 68 | ||
More Information
Set Membership: |
|

HSP90B1
Genomic Map