Assay Details
| Gene Symbol: ATP5PF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174717.3 | NP_777142.1 | 3 - 4 | 340 | 130 | |
| GenBank mRNA | BC103429.1 | - | 3 - 4 | 343 | 130 | |
| M19217.1 | - | 3 - 4 | 343 | 130 | ||
More Information
Set Membership: |
|

ATP5PF
Genomic Map