Assay Details
| Gene Symbol: RDH10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174734.2 | NP_777159.1 | 4 - 5 | 873 | 81 | |
| GenBank mRNA | AF456766.1 | - | 4 - 5 | 873 | 81 | |
| BC134557.1 | - | 4 - 5 | 814 | 81 | ||
More Information
Set Membership: |
|

RDH10
Genomic Map