Assay Details
| Gene Symbol: KIR2DL5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174740.2 | NP_777165.1 | 5 - 6 | 710 | 66 | |
| GenBank mRNA | AF490399.1 | - | 5 - 6 | 710 | 66 | |
| AY075102.1 | - | 5 - 6 | 710 | 66 | ||
More Information
Set Membership: |
|

KIR2DL5A
Genomic Map