Assay Details
| Gene Symbol: GNG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174811.4 | NP_777236.1 | 329 | 128 | ||
| GenBank mRNA | BC102572.1 | - | 330 | 128 | ||
| M95779.1 | - | 2 - 3 | 297 | 128 | ||
More Information
Set Membership: |
|

GNG5
Genomic Map