Assay Details
| Gene Symbol: SIRPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175788.1 | NP_786982.1 | 6 - 7 | 1306 | 65 | |
| GenBank mRNA | Y11045.1 | - | 1338 | 65 | ||
| Y11046.1 | - | 1331 | 65 | |||
More Information
Set Membership: |
|

SIRPA
Genomic Map