Assay Details
| Gene Symbol: NDUFA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175820.3 | NP_787014.1 | 2 - 3 | 216 | 73 | |
| GenBank mRNA | BC102882.1 | - | 2 - 3 | 216 | 73 | |
| X64897.1 | - | 206 | 73 | |||
More Information
Set Membership: |
|

NDUFA4
Genomic Map