Assay Details
| Gene Symbol: NDUFA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176672.2 | NP_788845.1 | 2 - 3 | 209 | 68 | |
| GenBank mRNA | AJ316011.1 | - | 2 - 3 | 209 | 68 | |
| BC102406.1 | - | 2 - 3 | 194 | 68 | ||
More Information
Set Membership: |
|

NDUFA13
Genomic Map