Assay Details
| Gene Symbol: CEBPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176784.2 | NP_789741.2 | 1 | 1153 | 124 | |
| GenBank mRNA | AY458599.1 | - | 1 | 1153 | 124 | |
| BC149006.1 | - | 2 | 658 | 124 | ||
More Information
Set Membership: |
|

CEBPA
Genomic Map