Assay Details
| Gene Symbol: GGTA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177511.2 | NP_803477.1 | 1 - 2 | 248 | 55 | |
| GenBank mRNA | J04989.1 | - | 1 - 2 | 248 | 55 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_177511.2 transcripts.
Set Membership: |
|

GGTA1
Genomic Map