Assay Details
| Gene Symbol: PRNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271625.3 | NP_001258554.1 | 1 - 2 | 74 | 82 | |
| NM_181015.4 | NP_851358.2 | 1 - 2 | 74 | 82 | ||
| GenBank mRNA | AB001468.1 | - | 1 - 2 | 53 | 82 | |
| BC119821.1 | - | 1 - 2 | 51 | 82 | ||
| D10612.1 | - | 1 - 2 | 53 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC119821.1, NM_001271625.3, NM_181015.4 transcripts.
Set Membership: |
|

PRNP
Genomic Map