Assay Details
| Gene Symbol: PNO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183085.1 | NP_898908.1 | 5 - 6 | 661 | 79 | |
| GenBank mRNA | AY336967.1 | - | 5 - 6 | 661 | 79 | |
| BC147981.1 | - | 5 - 6 | 663 | 79 | ||
More Information
Set Membership: |
|

PNO1
Genomic Map