Assay Details
| Gene Symbol: RAET1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205785.1 | NP_991354.1 | 1 - 2 | 136 | 112 | |
| GenBank mRNA | AY160681.1 | - | 1 - 2 | 136 | 112 | |
More Information
Set Membership: |
|

RAET1G
Genomic Map