Assay Details
Gene Symbol: RAET1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205785.1 | NP_991354.1 | 3 - 4 | 678 | 62 | |
GenBank mRNA | AY160681.1 | - | 3 - 4 | 678 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
