Assay Details
| Gene Symbol: COX4I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001439.3 | NP_001001439.1 | 1 - 2 | 350 | 91 | |
| GenBank mRNA | BC102733.1 | - | 1 - 2 | 351 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102733.1, NM_001001439.3 transcripts.
Set Membership: |
|

COX4I1
Genomic Map