Assay Details
Gene Symbol: ST3GAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002892.2 | NP_001002892.1 | 2 - 3 | 1024 | 70 | |
GenBank mRNA | AJ748841.1 | - | 2 - 3 | 536 | 70 | |
BC105252.1 | - | 2 - 3 | 1024 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
