Assay Details
| Gene Symbol: MYD88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014382.2 | NP_001014404.1 | 1 - 2 | 366 | 72 | |
| GenBank mRNA | AJ853453.1 | - | 1 - 2 | 333 | 72 | |
| BC102851.1 | - | 1 - 2 | 366 | 72 | ||
| KT601039.1 | - | 1 - 2 | 329 | 72 | ||
More Information
Set Membership: |
|

MYD88
Genomic Map