Assay Details
| Gene Symbol: RPL29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014862.2 | NP_001014862.1 | 2 - 3 | 83 | 71 | |
| GenBank mRNA | AY911327.1 | - | 2 - 3 | 87 | 71 | |
| BC102218.1 | - | 2 - 3 | 83 | 71 | ||
| BT021484.1 | - | 2 - 3 | 73 | 71 | ||
More Information
Set Membership: |
|

RPL29
Genomic Map