Assay Details
Gene Symbol: RPL29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014862.2 | NP_001014862.1 | 3 - 4 | 141 | 75 | |
GenBank mRNA | AY911327.1 | - | 3 - 4 | 145 | 75 | |
BC102218.1 | - | 3 - 4 | 141 | 75 | ||
BT021484.1 | - | 3 - 4 | 131 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
