Assay Details
Gene Symbol: TMEM171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014951.3 | NP_001014951.2 | 1 - 2 | 575 | 115 | |
GenBank mRNA | BC102968.1 | - | 1 - 2 | 576 | 115 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102968.1, NM_001014951.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
