Assay Details
Gene Symbol: TMEM171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014951.3 | NP_001014951.2 | 2 - 3 | 1281 | 134 | |
GenBank mRNA | BC102968.1 | - | 2 - 3 | 1282 | 134 | |
BT021523.1 | - | 2 - 3 | 795 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
