Assay Details
Gene Symbol: TMEM171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014951.3 | NP_001014951.2 | 3 - 4 | 1426 | 143 | |
GenBank mRNA | BC102968.1 | - | 3 - 4 | 1427 | 143 | |
BT021523.1 | - | 3 - 4 | 940 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
