Assay Details
| Gene Symbol: RPL26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015512.3 | NP_001015512.1 | 1 | 210 | 67 | |
| GenBank mRNA | AB098829.1 | - | 1 | 312 | 67 | |
| AB098945.1 | - | 1 | 188 | 67 | ||
| AY911370.1 | - | 2 - 3 | 212 | 67 | ||
| BC102057.1 | - | 1 | 211 | 67 | ||
| BC141993.1 | - | 1 | 199 | 67 | ||
More Information
Set Membership: |
|

RPL26
Genomic Map