Assay Details
| Gene Symbol: RPL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015556.2 | NP_001015556.1 | 1 | 223 | 53 | |
| GenBank mRNA | BC104507.1 | - | 1 | 223 | 53 | |
| BT021047.1 | - | 3 - 4 | 222 | 53 | ||
| DQ403039.1 | - | 2 - 3 | 131 | 53 | ||
More Information
Set Membership: |
|

RPL18
Genomic Map