Assay Details
| Gene Symbol: RNF114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024531.3 | NP_001019702.1 | 4 - 5 | 544 | 68 | |
| GenBank mRNA | BC102412.1 | - | 4 - 5 | 545 | 68 | |
| BT030581.1 | - | 4 - 5 | 512 | 68 | ||
| DQ010409.1 | - | 4 - 5 | 515 | 68 | ||
More Information
Set Membership: |
|

RNF114
Genomic Map