Assay Details
| Gene Symbol: MTG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025327.2 | NP_001020498.2 | 6 - 7 | 529 | 114 | |
| GenBank mRNA | BC103282.1 | - | 6 - 7 | 529 | 114 | |
| DQ058609.1 | - | 6 - 7 | 517 | 114 | ||
More Information
Set Membership: |
|

MTG1
Genomic Map