Assay Details
| Gene Symbol: RPL31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025341.3 | NP_001020512.1 | 22 | 51 | ||
| GenBank mRNA | AY911357.1 | - | 1 - 2 | 24 | 51 | |
| BC102125.1 | - | 25 | 51 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102125.1, NM_001025341.3 transcripts.
Set Membership: |
|

RPL31
Genomic Map