Assay Details
| Gene Symbol: RPL31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025341.3 | NP_001020512.1 | 369 | 66 | ||
| GenBank mRNA | AY911357.1 | - | 4 - 5 | 371 | 66 | |
| BC102125.1 | - | 372 | 66 | |||
More Information
Set Membership: |
|

RPL31
Genomic Map