Assay Details
| Gene Symbol: RPS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025342.2 | NP_001020513.1 | 1 | 49 | 112 | |
| GenBank mRNA | AY911350.1 | - | 1 - 2 | 52 | 112 | |
| BC102749.1 | - | 1 | 49 | 112 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102749.1, NM_001025342.2 transcripts.
Set Membership: |
|

RPS13
Genomic Map