Assay Details
| Gene Symbol: RPS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025342.2 | NP_001020513.1 | 1 | 179 | 78 | |
| GenBank mRNA | AY911350.1 | - | 3 - 4 | 182 | 78 | |
| BC102749.1 | - | 1 | 179 | 78 | ||
More Information
Set Membership: |
|

RPS13
Genomic Map