Assay Details
| Gene Symbol: RPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033613.2 | NP_001028785.1 | 3 - 4 | 293 | 130 | |
| GenBank mRNA | AB098951.1 | - | 2 - 3 | 202 | 130 | |
| AF013215.1 | - | 2 - 3 | 249 | 130 | ||
| BC102227.1 | - | 3 - 4 | 295 | 130 | ||
More Information
Set Membership: |
|

RPS2
Genomic Map