Assay Details
| Gene Symbol: RPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033613.2 | NP_001028785.1 | 7 - 8 | 823 | 65 | |
| GenBank mRNA | AF013215.1 | - | 6 - 7 | 779 | 65 | |
| BC102227.1 | - | 7 - 8 | 825 | 65 | ||
More Information
Set Membership: |
|

RPS2
Genomic Map