Assay Details
Gene Symbol: DNAJB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033763.1 | NP_001028935.1 | 2 - 3 | 833 | 151 | |
GenBank mRNA | BC104503.1 | - | 2 - 3 | 833 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
